Order Entry
France
ContactUsLinkComponent
Nicotinic acid (Niacin), Millipore®
Irritant
Nicotinic acid (Niacin), Millipore®
Catalog # 481918-100
CAS Number:  
Irritant
Nicotinic acid (Niacin), Millipore®
Catalog # 481918-100
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :123.11 g/mol
Merck Index:13,06552
Boiling Pt:258 °C (1013 hPa)
Melting Pt:142 °C
Density:1.473 g/cm³ (20 °C)
Flash Pt:193 °C
Storage Temperature:Ambient
EINECS:200-441-0
 :GHS07
CAS Number:59-67-6
Formula:C₆H₅NO₂
MDL Number:MFCD00006391