Ethyl nicotinate for synthesis, Sigma-Aldrich®
Catalog # 8.06754.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 ml
- Seq:2590
Specifications
Specification Test Results
- Solubility:50 g/l (20 °C)
- Melting Point:8 - 9 °C
- Molar Mass:151.16 g/mol
- Boiling Point:103 - 105 °C (7 hPa)
- Flash Point:93 °C
- Refractive Index:1.504 (20 °C, 589 nm)
- Density:1.11 g/cm3 (20 °C)
Specification Test Results
About this item
:
:151.16 g/mol
Boiling Pt:223 °C (1013 hPa)
Density:1.115 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:210-370-7
:GHS08
Storage Temperature:Ambient
CAS Number:614-18-6
Formula:C₈H₉NO₂
MDL Number:MFCD00006389