Mercury(II) chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS
Catalog # 25384.298
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay (dried basis):99.5 --> 100.5 %
- Identification:Passes test
- Acidity or alkalinity:Passes test
- Appearance of solution:Passes test
- Mercury (I) chloride:Passes test
- Solution in ethyl ether:Passes test
- Loss on drying (under vacuum; 24 h):Max. 1.0 %
- Residue after reduction:Max. 0.02 %
- Fe (Iron):Max. 20 ppm
- Conforms to Reag. Ph.Eur.:Passes test
- Conforms to ACS:Passes test
Specification Test Results
About this item
:
:271.5 g/mol
Merck Index:13,05901
Boiling Pt:302 °C (1013 hPa)
Melting Pt:273 °C
Density:5.44 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-299-8
UN:1624
ADR:6.1,II
:GHS08
:GHS06
:GHS05
:GHS09
CAS Number:7487-94-7
Formula:HgCl₂
MDL Number:MFCD00011041