Order Entry
France
ContactUsLinkComponent
Mercury(II) chloride ACS, Reag. Ph. Eur. for analysis, Supelco®
Mercury(II) chloride ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Mercury(II) chloride ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (complexometric):
    ≥ 99.5 %
  • Assay (complexometric; calculated on dried substance):
    99.5 - 100.5 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • Solution in Ether:
    passes test
  • Acidity or alkalinity:
    passes test
  • Mercury(I)-chloride:
    passes test
  • Fe (Iron):
    ≤ 0.002 %
  • Residue after reduction:
    ≤ 0.02 %
  • Loss on drying (24 h im Vakuum):
    ≤ 1.0 %

Specification Test Results

Related Information

Frequently Bought Together