Mercury(II) chloride ≥99.5% (dried basis), Puriss. p.a. ACS, ISO, Fluka™
Catalog # 31005-500G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay (calculated to the dried substance):Min. 99.5 %
- Loss on drying (on P₂O₅):Max. 1.0 %
- Residue on ignition a. reducing with HCOOH (SO₄)*:Max. 0.02 %
- Water insoluble matter:Max. 0.01 %
- Silver (Ag):Max. 0.0005 %
- Calcium (Ca):Max. 0.001 %
- Cadmium (Cd):Max. 0.0005 %
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 0.001 %
- Iron (Fe):Max. 0.0005 %
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 0.001 %
- Manganese (Mn):Max. 5 ppm
- Sodium (Na):Max. 0.005 %
- Nickel (Ni):Max. 0.0005 %
- Lead (Pb):Max. 0.001 %
- Zinc (Zn):Max. 0.0005 %
- Other heavy metals (as Pb):Max. 0.001 %
- Mercury(I) chloride:Max. 0.05 %
- Total S (as SO₄):Max. 50 ppm
- Sulfate (SO₄):Max. 0.02 %
- Total N:Max. 0.002 %
- In ether insoluble matter:complying
- Appearance of the solution:complying
Specification Test Results
About this item
:
:271.5 g/mol
Merck Index:13,05901
Boiling Pt:302 °C
Melting Pt:276 °C
Density:5.44 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-299-8
UN:1624
ADR:6.1,II
:GHS06
:GHS08
:GHS05
:GHS09
CAS Number:7487-94-7
Formula:HgCl₂
MDL Number:MFCD00011041