

Copper(II) oxide ≥99.9995% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1317-38-0
:
Formula:CuO
MDL Number:MFCD00010979
:79.55 g/mol
Merck Index:14,2646
Boiling Pt:2000 °C (1013 hPa)
Melting Pt:1326 °C
Density:6.3
Storage Temperature:Ambient
EINECS:215-269-1
UN:3077
ADR:9,III
:GHS07
:GHS09
Related Information
Frequently Bought Together