Order Entry
France
Orders LinkContactUsLinkComponent
 

Copper(II) oxide ≥99.7% (metals basis)

Irritant
Environment
Copper(II) oxide ≥99.7% (metals basis)
Certificates
Copper(II) oxide ≥99.7% (metals basis)
Copper(II) oxide ≥99.7% (metals basis)

3 Options Available Below

Product Details & Documents

Purity: 99.7% CAS No.: 1317-38-0 Molecular Formula: CuO Formula Weight: 79.55 Physical Form: -200 Mesh Powder Density: 6.3 Refractive Index: 2.63 Appearance: Black Odour: Odourless Melting Point: 1326° Merck Reference: 14,2646 UN No.: 3077 MDL No.: MFCD00010979

Synonyms:  Cupric oxide, Copper oxide, Copper monoxide granulated
  • CAS Number:1317-38-0
  • Formula:CuO
  • MDL Number:MFCD00010979
  • Molecular Weight:79.55 g/mol
  • Merck Index:14,2646
  • Boiling Pt:2000 °C (1013 hPa)
  • Melting Pt:1326 °C
  • Density:6.3
  • Storage Temperature:Ambient
  • EINECS:215-269-1
  • UN:3077
  • ADR:9,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.