Order Entry
France
ContactUsLinkComponent
Copper(II) bromide for synthesis, Sigma-Aldrich®
Corrosive
Irritant
Environment
Copper(II) bromide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
Environment
Copper(II) bromide for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    1200 g/l (25 °C)
  • Melting Point:
    498 °C
  • Molar Mass:
    223.35 g/mol
  • Bulk Density:
    2000 kg/m3
  • Boiling Point:
    900 °C (1013 hPa)
  • Density:
    4.71 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together