Order Entry
France
ContactUsLinkComponent
Ammonium thiocyanate ≥98%, purified
Irritant
Ammonium thiocyanate ≥98%, purified
Catalog # 21342.297
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Ammonium thiocyanate ≥98%, purified
Catalog # 21342.297
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Fe (Iron):
    Max. 5 ppm

Specification Test Results

About this item

 :
 :76.12 g/mol
Merck Index:13,00564
Melting Pt:150 °C
Density:1.305 g/cm³ (20 °C)
EINECS:217-175-6
 :GHS07
Storage Temperature:Ambient
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428