Order Entry
France
ContactUsLinkComponent
Ammonium thiocyanate 99+%, extra pure
Corrosive
Irritant
Ammonium thiocyanate 99+%, extra pure
Certificates
undefined
Corrosive
Irritant
Ammonium thiocyanate 99+%, extra pure

Specification Test Results

  • Appearance:
    White Crystals
  • Titration Argentometric:
    ≥99.0 % (on dried basis)
  • Water:
    ≤1.0 %
  • pH:
    4.0 to 6.0 (5 % in water)
  • Iron (Fe):
    ≤2 ppm

Specification Test Results

Related Information

Frequently Bought Together