Order Entry
France
ContactUsLinkComponent
Ammonium dihydrogen phosphate ≥97.5%, purified
Ammonium dihydrogen phosphate ≥97.5%, purified
Catalog # 21303.293
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium dihydrogen phosphate ≥97.5%, purified
Catalog # 21303.293
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 97.5 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Fe (Iron):
    Max. 50 ppm

Specification Test Results

About this item

 :
Merck Index:13,00547
Melting Pt:190 °C
Density:1.8 g/cm³ (20 °C)
 :115.03 g/mol
Storage Temperature:Ambient
EINECS:231-764-5
REACH:01-2119488166-29
CAS Number:7722-76-1
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396