Lead(II) sulphate, Reagent Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7446-14-2
:
Formula:O₄PbS
MDL Number:MFCD00011166
:303.26 g/mol
Merck Index:14,5420
Melting Pt:1170 °C
Density:6.2
EINECS:231-198-9
UN:2291
ADR:6.1,III
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together