Lead(II) sulphate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7446-14-2
:
Formula:O₄PbS
MDL Number:MFCD00011166
:303.26 g/mol
Merck Index:12,05444
Melting Pt:1170 °C
Density:6,0631 g/cm³ (18 °C)
EINECS:231-198-9
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Identification:Confirmed
- Elemental Analysis:≥98.5 % (AES)
Specification Test Results
Frequently Bought Together