Order Entry
France
Orders LinkContactUsLinkComponent
Aluminium oxide, regular drying agent

Aluminium oxide, regular drying agent

Aluminium oxide, regular drying agent
Certificates
Aluminium oxide, regular drying agent
Aluminium oxide, regular drying agent

3 Options Available Below

Product Details & Documents

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: 3.2mm Spheres Odour: Odourless MDL No.: MFCD00003424

Dry gases and liquids. Gases include: CO2, acetylene, air, Ar, ethylene, He, H2, N2, O2. Liquids include: benzene, hexane, oil, pentane.

Synonyms:  Aluminium(III) oxide, Alumina C, Aluminum oxide
  • CAS Number:1344-28-1
  • Formula:Al₂O₃
  • MDL Number:MFCD00003424
  • Molecular Weight:101.96 g/mol
  • Merck Index:13,00355
  • Boiling Pt:2980 °C (1013 hPa)
  • Melting Pt:2045 °C
  • Density:3.94 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-691-6
  • REACH:01-2119529248-35
Product Family Options

Product Information