Aluminium oxide, intermediate surface area catalyst support
Supplier: Thermo Fisher Scientific
Aluminium oxide, intermediate surface area catalyst support
Supplier: Thermo Fisher Scientific
Certificates
Jump to:
About this item
Physical Form: 1/8" pellets MDL No.: MFCD00003424
Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide
- CAS Number:1344-28-1
- Formula:Al₂O₃
- MDL Number:MFCD00003424
- Molecular Weight:101.96 g/mol
- Merck Index:13,00355
- Boiling Pt:2980 °C (1013 hPa)
- Melting Pt:2045 °C
- Density:3.94 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-691-6
- REACH:01-2119529248-35
2 Options Available Below
Product Details & Documents
Physical Form: 1/8" pellets MDL No.: MFCD00003424
Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide
- CAS Number:1344-28-1
- Formula:Al₂O₃
- MDL Number:MFCD00003424
- Molecular Weight:101.96 g/mol
- Merck Index:13,00355
- Boiling Pt:2980 °C (1013 hPa)
- Melting Pt:2045 °C
- Density:3.94 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-691-6
- REACH:01-2119529248-35
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



