Order Entry
France
ContactUsLinkComponent
Calcium sulphate, anhydrous 99%, pure
Calcium sulphate, anhydrous 99%, pure
Certificates
undefined
Calcium sulphate, anhydrous 99%, pure

Specification Test Results

  • Appearance:
    Off-white Crystalline powder
  • Assay:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together