Order Entry
France
ContactUsLinkComponent
 
 
 
Calcium hydroxide, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
 
 
 
Calcium hydroxide, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
 

 

  • Solubility:
    1.7 g/l (20 °C)
  • Melting Point:
    550 °C (decomposition)
  • Molar Mass:
    74.09 g/mol
  • Bulk Density:
    400 kg/m3
  • Boiling Point:
    2850 °C (1013 hPa) (decomposition)
  • Vapor Pressure:
    (20 °C)
  • Density:
    2.24 g/cm3 (20 °C)
  • pH:
    12.6 (H2O, 20 °C) (saturated solution)

 

Frequently Bought Together