Sodium sulphite, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 1777.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:min. 95 %
- Chloride (Cl):max. 0.02 %
- Iron (Fe):max. 0.002 %
- Heavy Metals (as Pb):max. 0.005 %
- Trace Impurities (in ppm)Arsenic (As):max. 2
Specification Test Results
About this item
:
:126.04 g/mol
Merck Index:13,08757
Melting Pt:> 500 °C
Density:2.63 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-821-4
REACH:01-2119537420-49
CAS Number:7757-83-7
Formula:Na₂SO₃
MDL Number:MFCD00003503