Sodium dihydrogen phosphate hydrate ACS, Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:10049-21-5
:
Formula:NaH₂PO₄·xH₂O
MDL Number:MFCD00149208
Merck Index:12,08806
Melting Pt:∼650 °C
Density:0.9996 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
Specification Test Results
- Form:White Solid
- Purity:≥98% by assay
- Contaminants:Heavy metals: ≤0.001%
Specification Test Results
Frequently Bought Together