Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lithium chloride
Lithium chloride
Lithium chloride
PRLS87618.440
:
PROLABO CMP
:
Lithium chloride
PRLS87618.440
:
PROLABO CMP
:
Pk:
25 L
Seq:
5327
:
:
42.39 g/mol
:
13,05550
:
1382 °C (1013 hPa)
:
614 °C
:
2.07 g/cm³ (20 °C)
:
Ambient
:
231-212-3
:
GHS07
:
7447-41-8
:
LiCl
:
MFCD00011078