Octamethylcyclotetrasiloxane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:556-67-2
:
Formula:[-Si(CH₃)₂O-]₄
MDL Number:MFCD00003269
:296.62 g/mol
Merck Index:12,06843
Boiling Pt:175 °C (1013 hPa)
Melting Pt:18 °C
Density:0.95 g/cm³ (20 °C)
EINECS:209-136-7
UN:1993
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:51 °C
:GHS02
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:- 0.001 g/l (25 °C)
- Melting Point:18 °C
- Molar Mass:296.62 g/mol
- Boiling Point:176 °C
- Vapor Pressure:1.3 hPa (20 °C)
- Flash Point:51 °C
- Explosion Limit:0.4 - 11.7 %(V)
- Density:0.95 g/cm3
- Ignition Point:400 °C
Specification Test Results
Frequently Bought Together