myo-Inositol, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:87-89-8
:
Formula:C₆H₁₂O₆
MDL Number:MFCD00077932
:180.16 g/mol
Merck Index:13,05001
Boiling Pt:291 °C (1013 hPa)
Melting Pt:210 °C
Density:1.752 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:201-781-2
Frequently Bought Together