Order Entry
France
ContactUsLinkComponent
myo-Inositol
myo-Inositol
Catalog # A1716.0250
Supplier:  PanReac AppliChem
CAS Number:  
myo-Inositol
Catalog # A1716.0250
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :180.16 g/mol
Merck Index:13,05001
Boiling Pt:291 °C (1013 hPa)
Melting Pt:210 °C
Density:1.752 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-781-2
CAS Number:87-89-8
Formula:C₆H₁₂O₆
MDL Number:MFCD00077932