Order Entry
France
ContactUsLinkComponent
2-Phenylethyl isothiocyanate
2-Phenylethyl isothiocyanate
Catalog # MOLE24234941-5G
Supplier:  Molekula
CAS Number:  
2-Phenylethyl isothiocyanate
Catalog # MOLE24234941-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :163.24 g/mol
Density:1.09 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:218-855-5
CAS Number:2257-09-2
Formula:C₆H₅CH₂CH₂NCS
MDL Number:MFCD00004821