

2-Phenylethyl isothiocyanate 96%
:2257-09-2
:
:C₆H₅CH₂CH₂NCS
:MFCD00004821
:163.24 g/mol
:113 °C (1 mmHg)
:1.1 g/cm³
:100 °C
:218-855-5
:2810
:GHS07
:GHS08
:Ambient
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5860 to 1.5910 (20°C, 589 nm)
- GC:≥95 %
Frequently Bought Together