Order Entry
France
ContactUsLinkComponent
 
3-Fluorobenzylamine 97%
 
undefined
3-Fluorobenzylamine 97%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear colorless to yellow Liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5125 to 1.5175 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • NMR (1H-NMR):
    Consistent with structure

 

 

Frequently Bought Together