2-Chloro-4-(trifluoromethyl)aniline
Supplier: Thermo Scientific
Certificates
About this item
Synonyms: 4-Amino-3-chloro-α,α,α-trifluorotoluene, 2-Chloro-α,α,α-trifluoro-p-toluidine, 4-Amino-3-chlorobenzotrifluoride
CAS Number:39885-50-2
:
Formula:C₇H₅ClF₃N
MDL Number:MFCD00042563
:195.57 g/mol
Boiling Pt:89 °C (0.5 mmHg)
EINECS:254-674-8
UN:2810
ADR:6.1,III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together