trans-Stilbene for synthesis, Sigma-Aldrich®
Catalog # 8.14152.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:0.00029 g/l (25 °C)
- Melting Point:121 - 124 °C
- Molar Mass:180.24 g/mol
- Boiling Point:306 - 307 °C (1013 hPa)
- Density:1.14 g/cm3 (20 °C)
Specification Test Results
About this item
Synonyms: trans-1,2-Diphenylethylene, (E)-1,2-Diphenylethene, trans-1,2-Diphenylethene
:
:180.25 g/mol
Merck Index:12,08972
Boiling Pt:306 °C (1013 hPa)
Melting Pt:121…124 °C
Density:0.9707 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:203-098-5
CAS Number:103-30-0
Formula:C₁₄H₁₂
MDL Number:MFCD00064300