

Benzoic anhydride 98%
:93-97-0
:
:C₁₄H₁₀O₃
:MFCD00003073
:13,01093
:360 °C (1013 hPa)
:38…44 °C
:1.19 g/cm³
:>112 °C
:226.23 g/mol
:Ambient
:202-291-1
:GHS05
:GHS07
- Appearance:White to off-white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:38 to 44 °C
- HPLC:≥97.5 %
Frequently Bought Together