Phthalic acid 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Pathalic acid
- CAS Number:88-99-3
- Formula:C₈H₆O₄
- MDL Number:MFCD00002467
- Molecular Weight:166.13 g/mol
- Merck Index:12,07527
- Boiling Pt:315 - 317 °C (1013 hPa)
- Melting Pt:200…205 °C (dec.)
- Density:1,576 g/cm³ (20 °C)
- Flash Pt:168 °C
- Storage Temperature:Ambient
- EINECS:201-873-2
3 Options Available Below
Product Details & Documents
Synonyms: Pathalic acid
- CAS Number:88-99-3
- Formula:C₈H₆O₄
- MDL Number:MFCD00002467
- Molecular Weight:166.13 g/mol
- Merck Index:12,07527
- Boiling Pt:315 - 317 °C (1013 hPa)
- Melting Pt:200…205 °C (dec.)
- Density:1,576 g/cm³ (20 °C)
- Flash Pt:168 °C
- Storage Temperature:Ambient
- EINECS:201-873-2
Documents
ACRO13107
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:200 to 205 °C (Dec)
- Titration with NaOH:≥98.5 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤50 ppm
- Heavy metals (as Pb):≤10 ppm
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



