O-Acetylsalicylic acid
Catalog # EHERC10024000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-Acetoxybenzoic acid, Acetylsalicylic acid, Aspirin, Aspirin®
:
:180.16 g/mol
Merck Index:13,00856
Boiling Pt:140 °C (1013 hPa)
Melting Pt:138…140 °C
Density:1.39 g/cm³ (20 °C)
Flash Pt:250 °C
Storage Temperature:Ambient
EINECS:200-064-1
:GHS07
CAS Number:50-78-2
Formula:2-(CH₃CO₂)C₆H₄CO₂H
MDL Number:MFCD00002430