
5-Fluoroisatin
Catalog # MOLE27999310-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:165.12 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:443-69-6
Formula:C₈H₄FNO₂
MDL Number:MFCD00022795