Order Entry
France
ContactUsLinkComponent
 
 
 
 
Piperidine ≥99.5%, Biotechnology Grade, Burdick & Jackson™
 
 
 
 
Piperidine ≥99.5%, Biotechnology Grade, Burdick & Jackson™

 

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.50 %
  • Refractive index (n 20/D):
    1.451 - 1.455
  • Water (Karl Fischer):
    Max. 0.3 %
  • Absorbance at 370 nm:
    Max. 0.050
  • Absorbance at 290 nm:
    Max. 1.000
  • Refractive Index:
    n20/D 1.452 (lit.)
  • Vapor Density:
    3 (vs air)

 

 

Frequently Bought Together