Order Entry
France
ContactUsLinkComponent
Piperidine for synthesis, Sigma-Aldrich®
Piperidine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Piperidine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) miscible in all proportions
  • Melting Point:
    -10 °C
  • Molar Mass:
    85.15 g/mol
  • Boiling Point:
    106 °C (1013 hPa)
  • Vapor Pressure:
    34 hPa (20 °C)
  • Flash Point:
    16 °C
  • Explosion Limit:
    1.5 - 10.3 %(V)
  • Density:
    0.86 g/cm3 (20 °C)
  • pH:
    12.6 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    320 °C
  • Saturation concentration:
    115 g/m3 (20 °C) Air
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive material

Specification Test Results

Related Information

Frequently Bought Together