Order Entry
France
Orders LinkContactUsLinkComponent
 

N-Bromosuccinimide 99%

Oxidizing
Corrosive
Irritant
Environment
N-Bromosuccinimide 99%
Certificates
N-Bromosuccinimide 99%

5 Options Available Below

Product Details & Documents
Synonyms:  NBS, 1-Bromopyrrolidine-2,5-dione
  • CAS Number:128-08-5
  • Formula:C₄H₄BrNO₂
  • MDL Number:MFCD00005510
  • Molecular Weight:177.99 g/mol
  • Merck Index:13,01423
  • Boiling Pt:339 °C (1013 hPa)
  • Melting Pt:175…178 °C (dec.)
  • Density:2,098 g/cm³ (20 °C)
  • Flash Pt:180 °C
  • Storage Temperature:Refrigerator
  • EINECS:204-877-2
  • UN:3084
  • ADR:8,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Iodometric:
    98.5 to 101.5 % (on dry substance)
  • Loss on drying:
    ≤1 %
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.