
3-Cyclohexylpropyne ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17715-00-3
:
Formula:C₉H₁₄
MDL Number:MFCD00041562
:122.21 g/mol
Boiling Pt:157…158 °C
Density:0.845
Flash Pt:35 °C (95 °F)
Storage Temperature:Ambient
UN:3295
ADR:3,III
:GHS02
Related Information
Frequently Bought Together