Order Entry
France
ContactUsLinkComponent
Citraconic anhydride for synthesis, Sigma-Aldrich®
Corrosive
Health
Citraconic anhydride for synthesis, Sigma-Aldrich®
Catalog # 8.41321.0100
Supplier:  Merck
CAS Number:  
Corrosive
Health
Citraconic anhydride for synthesis, Sigma-Aldrich®
Catalog # 8.41321.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Melting Point:
    7 - 8 °C
  • Molar Mass:
    112.08 g/mol
  • Boiling Point:
    213 - 214 °C (1013 hPa)
  • Vapor Pressure:
    1.3 hPa (47 °C)
  • Flash Point:
    101 °C
  • Density:
    1.243 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :112.08 g/mol
Boiling Pt:213 °C (1013 hPa)
Melting Pt:7…8 °C
Density:1.243 g/cm³ (20 °C)
Flash Pt:101 °C
Storage Temperature:Refrigerator
EINECS:210-459-0
UN:3265
ADR:8,II
 :M-Clarity™ quality level = MQ200
 :GHS05
 :GHS08
CAS Number:616-02-4
Formula:C₅H₄O₃
MDL Number:MFCD00005522