


Citraconic anhydride 98%
:616-02-4
:
:C₅H₄O₃
:MFCD00005522
:112.08 g/mol
:213 °C (760 mmHg)
:6…7 °C
:1.245 g/cm³
:101 °C
:Ambient
:210-459-0
:2922
:8,III
:GHS06
:GHS08
:GHS05
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4690 to 1.4730 (20°C, 589 nm)
- Titration with NaOH:97.5 to 102.5 %
- NMR (1H-NMR):Consistent with structure
Frequently Bought Together