
Fumaric acid 99+%
:110-17-8
:
:C₄H₄O₄
:MFCD00002700
:116.07 g/mol
:13,04308
:356 °C (1013 hPa)
:295…300 °C (subl.)
:1.62 g/cm³
:230 °C
:203-743-0
:GHS07
:Ambient
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:295 to 300 °C
- Water:≤0.25 % (K.F.)
- HPLC:≥99.0 %
- Titration with NaOH:99.0 to 101.0 % (On anhydrous substance)
Frequently Bought Together