Order Entry
France
ContactUsLinkComponent
Dimethyl fumarate
Dimethyl fumarate
Catalog # EHERC13955800
Supplier:  EHRENSTORFER
CAS Number:  
Dimethyl fumarate
Catalog # EHERC13955800
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :144.13 g/mol
Merck Index:13,04308
Boiling Pt:192 °C (1013 hPa)
Melting Pt:102 °C
Density:1.37 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-849-0
CAS Number:624-49-7
Formula:C₆H₈O₄
MDL Number:MFCD00064438