Diethyl fumarate 98%
:623-91-6
:
:C₈H₁₂O₄
:MFCD00064455
:172.18 g/mol
:218…219 °C (760 mmHg)
:1…2 °C
:1.05 g/cm³
:91 °C
:210-819-7
:GHS07
:Ambient
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4396 to 1.4416 (20°C, 589 nm)
- GC:≥97.5 %
Frequently Bought Together