HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid)
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7365-45-9
:
Formula:C₈H₁₈N₂O₄S
MDL Number:MFCD00006158
Merck Index:13,04672
Melting Pt:210…215 °C
Density:1.448 g/cm³ (20 °C)
:238.31 g/mol
Storage Temperature:Ambient
EINECS:230-907-9
Frequently Bought Together