HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% for biochemistry
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7365-45-9
:
Formula:C₈H₁₈N₂O₄S
MDL Number:MFCD00006158
Merck Index:13,04672
Melting Pt:210…215 °C
Density:1.448 g/cm³ (20 °C)
:238.31 g/mol
Storage Temperature:Ambient
EINECS:230-907-9
Specification Test Results
- Assay:Min. 99.5 %
- Appearance:White powder
- Appearance of solution (1 mol/l):Clear colourless liquid
- IR Spectrum:Passes test
- pH (5%; 25°C):5.0 - 6.5
- Ignition residue:Max. 0.3 %
- LOD (140°C):Max. 0.5 %
- Cl (Chloride):Max. 300 ppm
- PO4 (Phosphate):Max. 10 ppm
- SO4 (Sulphate):Max. 100 ppm
- Al (Aluminium):Max. 5 ppm
- Cu (Copper):Max. 5 ppm
- Fe (Iron):Max. 5 ppm
- Na (Sodium):Max. 0.1 %
- Pb (Lead):Max. 5 ppm
- Absorbance (260 nm) (1 mol/l):Max. 0.10
- Absorbance (280 nm) (1 mol/l):Max. 0.10
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Plastic bucket for solids UN Y (ZPACK13247010) 2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
Avantor Cell Culture Solutions.pdf
Product Literature
spec_VWRC441485H spec_VWRC441485H spec_VWRC441487M spec_VWRC441487M spec_VWRC44148BC
Product specification
Frequently Bought Together