Order Entry
France
ContactUsLinkComponent
 
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) free acid ≥99%, high purity
 
undefined
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) free acid ≥99%, high purity
 

 

  • DNase:
    none detected
  • Heavy Metals (as Pb):
    ≤0.0005%
  • Iron:
    ≤0.0005%
  • pKa @20 °C:
    7.45 – 7.65
  • Protease:
    none detected
  • Purity (Dry Basis):
    >99%
  • Residue on Ignition:
    <0.2%
  • RNase:
    none detected

 

 

Frequently Bought Together