
(3,4-Dihydroxyphenyl)acetic acid 98%
:102-32-9
:
:(C₆H₃(OH)₂)CH₂COOH
:MFCD00004338
:168.15 g/mol
:337 °C (1013 hPa)
:126…130 °C
:Ambient
:203-024-1
:GHS07
- Appearance:Beige to light brown to purple-grey Powder
- Infrared spectrum:Conforms
- Melting point:127 to 132 °C
- HPLC:≥97.5 %
Frequently Bought Together