Order Entry
France
ContactUsLinkComponent
Benzilic acid for synthesis, Sigma-Aldrich®
Benzilic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Benzilic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    1.41 g/l (25 °C) (experimental)
  • Melting Point:
    150 - 152 °C
  • Molar Mass:
    228.24 g/mol
  • Boiling Point:
    180 °C (17.3 hPa)
  • Ignition Point:
    530 °C

Specification Test Results

Related Information

Frequently Bought Together