Order Entry
France
ContactUsLinkComponent
(3,4-Dimethoxyphenyl)acetic acid
(3,4-Dimethoxyphenyl)acetic acid
  MOLEM80298319
 :  Molekula
 :  
(3,4-Dimethoxyphenyl)acetic acid
  MOLEM80298319
 :  Molekula
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
 :
 :196.2 g/mol
 :327 °C (1013 hPa)
 :97…99 °C
 :Ambient
 :202-244-5
 :93-40-3
 :(C₆H₃(OCH₃)₂)CH₂COOH
 :MFCD00004335