

tert-Butanol, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:75-65-0
:
Formula:(CH₃)₃COH
MDL Number:MFCD00004464
:74.12 g/mol
Merck Index:12,01577
Boiling Pt:82.2 °C (1013 hPa)
Melting Pt:25.3 °C
Density:0.7887 g/cm³ (20 °C)
EINECS:200-889-7
UN:1120
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:11 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together