

Pivalic anhydride 99%
:1538-75-6
:
:C₁₀H₁₈O₃
:MFCD00008842
:186.25 g/mol
:193 °C (760 mmHg)
:0.91 g/cm³
:57 °C
:Ambient
:216-263-1
:2920
:8,II
:GHS02
:GHS05
- Appearance:Clear colorless to light yellow liquid Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4080 to 1.4100 (20°C, 589 nm)
- GC:≥98.5 %
Frequently Bought Together