2-Mercaptoethanol ≥99% for biotechnology
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:60-24-2
:
Formula:HSCH₂CH₂OH
MDL Number:MFCD00004890
Merck Index:13,05894
Boiling Pt:157 °C (1013 hPa)
Melting Pt:–100 °C
Density:1.12 g/cm³ (20 °C)
Flash Pt:68 °C
Storage Temperature:Ambient
UN:2966
ADR:6.1,II
:78.14 g/mol
EINECS:200-464-6
:GHS06
:GHS05
:GHS09
Specification Test Results
- DNase:none detected
- Protease:none
- Purity:>99%
- RNase:none detected
- Water:≤0.5%
Specification Test Results
Related Information
Certificates
Search by Lot Number0482_2-MERCAPTOETHANOL BIOTECHNOLOGY GRADE_BSE_REV0.pdf
BSE/TSE Certificates
sds_VWRC0482
SDS
Frequently Bought Together